Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080437.1 | NP_001073906.1 | 25 | 3641 | ||
XM_011510931.2 | XP_011509233.1 | 26 | 3955 | |||
XM_011510932.2 | XP_011509234.1 | 26 | 3957 | |||
XM_011510933.2 | XP_011509235.1 | 26 | 3961 | |||
XM_011510934.2 | XP_011509236.1 | 26 | 3961 | |||
XM_017003773.1 | XP_016859262.1 | 26 | 3957 | |||
XM_017003774.1 | XP_016859263.1 | 25 | 3726 | |||
XM_017003775.1 | XP_016859264.1 | 26 | 3959 | |||
XM_017003776.1 | XP_016859265.1 | 25 | 3843 | |||
XM_017003777.1 | XP_016859266.1 | 24 | 3734 | |||
XM_017003778.1 | XP_016859267.1 | 25 | 3721 | |||
XM_017003779.1 | XP_016859268.1 | 26 | 3657 | |||
XM_017003780.1 | XP_016859269.1 | 25 | 3513 | |||
XM_017003781.1 | XP_016859270.1 | 25 | 3503 | |||
XM_017003782.1 | XP_016859271.1 | 21 | 2852 | |||
GenBank mRNA | AF439717.1 | - | 19 | 2484 | ||
AF439718.1 | - | 5 | 830 | |||
AK074062.1 | - | 24 | 3249 | |||
AK074075.1 | - | 7 | 1151 | |||
AL833363.1 | - | 1 | 222 | |||
BC027939.1 | - | 8 | 1479 |