Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZZZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308237.1 | NP_001295166.1 | 14 | 1648 | ||
NM_015534.5 | NP_056349.1 | 15 | 3112 | |||
XM_005270725.3 | XP_005270782.1 | 15 | 3187 | |||
XM_005270726.3 | XP_005270783.1 | 14 | 3088 | |||
XM_005270727.3 | XP_005270784.1 | 15 | 3184 | |||
XM_005270729.4 | XP_005270786.1 | 13 | 1598 | |||
XM_017000970.1 | XP_016856459.1 | 14 | 3085 | |||
XM_017000972.1 | XP_016856461.1 | 14 | 1616 | |||
GenBank mRNA | AK001224.1 | - | 5 | 485 | ||
AK074119.1 | - | 10 | 3527 | |||
AL080063.1 | - | 7 | 676 | |||
BC035079.2 | - | 14 | 1640 | |||
BC035397.1 | - | 11 | 1780 | |||
BC035818.2 | - | 11 | 2687 | 5554765 | ||
BX640658.1 | - | 11 | 1200 | |||
BX641001.1 | - | 15 | 3113 | |||
BX641096.1 | - | 15 | 3096 |