Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 3 | 370 | ||
NM_001271869.1 | NP_001258798.1 | 3 | 367 | |||
NR_073501.1 | 4 | 441 | ||||
NR_073502.1 | 2 | 320 | ||||
NR_073503.1 | 2 | 317 | ||||
GenBank mRNA | AB097042.1 | - | 2 | 108 | ||
AB097043.1 | - | 2 | 108 | |||
AB097044.1 | - | 2 | 99 | |||
AB097045.1 | - | 2 | 99 | |||
AB097046.1 | - | 3 | 158 | |||
AJ295987.1 | - | 2 | 104 | |||
AK297926.1 | - | 3 | 158 | |||
AK298576.1 | - | 2 | 104 | |||
AK316205.1 | - | 2 | 106 | |||
AL050028.1 | - | 2 | 104 | |||
BC023988.1 | - | 2 | 125 | 3825796 | ||
BC065538.1 | - | 2 | 105 | 6053254 | ||
BX640985.1 | - | 2 | 120 | |||
DA010145.1 | - | 3 | 162 | |||
DA438597.1 | - | 4 | 228 | |||
DB501645.1 | - | 3 | 309 | |||
DC320437.1 | - | 2 | 322 |