Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015617.2 | NP_056432.1 | 3 | 1060 | ||
XM_005254300.4 | XP_005254357.1 | 3 | 1181 | |||
XM_011521446.2 | XP_011519748.1 | 3 | 1455 | |||
GenBank mRNA | AF457207.1 | - | 3 | 1060 | ||
AL049925.1 | - | 1 | 154 | |||
EF625686.1 | - | 4 | 1185 |