Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012384.4 | NP_036516.1 | 7, 8 | 892 | ||
XM_005260202.4 | XP_005260259.1 | 7, 8 | 1103 | |||
XM_006723776.2 | XP_006723839.1 | 5, 6 | 544 | |||
XM_011528779.2 | XP_011527081.1 | 5, 6 | 608 | |||
GenBank mRNA | AB033095.1 | - | 7, 8 | 769 | ||
AF173867.1 | - | 6, 7 | 1168 | |||
AL133646.1 | - | 5, 6 | 616 | |||
BC036305.1 | - | 7, 8 | 819 | 5166938 | ||
BQ954281.1 | - | 6, 7 | 594 |