Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012139.3 | NP_036271.1 | 10 | 1184 | ||
NR_104040.1 | 11 | 1269 | ||||
NR_104041.1 | 9 | 1017 | ||||
GenBank mRNA | AJ243950.1 | - | 10 | 1050 | ||
AJ243951.1 | - | 9 | 883 | |||
AK292286.1 | - | 10 | 1184 | |||
AK294022.1 | - | 9 | 882 | |||
BC000707.2 | - | 11 | 1143 | |||
BC065375.1 | - | 10 | 1164 | 6672338 |