Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002053.2 | NP_002044.2 | 7 | 1317 | ||
GenBank mRNA | AB208912.1 | - | 6 | 1882 | ||
AK291783.1 | - | 7 | 1165 | |||
AK298032.1 | - | 4 | 491 | |||
AK303893.1 | - | 7 | 1120 | |||
AY920436.1 | - | 6 | 1101 | |||
BC002666.2 | - | 7 | 1317 | 3606865 | ||
BT006847.1 | - | 6 | 1097 | |||
M55542.1 | - | 7 | 1165 |