Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304485.1 | NP_001291414.1 | 5 | 542 | ||
NM_001304486.1 | NP_001291415.1 | 4 | 420 | |||
NM_001304487.1 | NP_001291416.1 | 4 | 454 | |||
NM_012460.3 | NP_036592.1 | 6 | 731 | |||
NR_130750.1 | 5 | 666 | ||||
NR_130752.1 | 3 | 355 | ||||
NR_130753.1 | 3 | 347 | ||||
GenBank mRNA | AF150100.1 | - | 5 | 422 | ||
AF152353.1 | - | 3 | 205 | |||
AK000666.1 | - | 5 | 440 | |||
AK312095.1 | - | 4 | 366 | |||
BC020213.1 | - | 6 | 631 | 4640518 | ||
BC054875.1 | - | 4 | 344 | 6663143 | ||
BF034731.1 | - | 5 | 386 | |||
BM811095.1 | - | 5 | 578 | |||
DB110327.1 | - | 3 | 284 | |||
HY139570.1 | - | 3 | 269 |