Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10G2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005466.2 | NP_001005466.2 | 1 | 520 | ||
GenBank mRNA | BC137371.1 | - | 1 | 520 | 9021369 | |
BC137382.1 | - | 1 | 520 | 9021380 |