Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012453.2 | NP_036585.1 | 2 | 311 | ||
XM_006715923.3 | XP_006715986.1 | 3 | 553 | |||
XM_011516027.2 | XP_011514329.1 | 2 | 204 | |||
GenBank mRNA | AF056183.1 | - | 2 | 139 | ||
AF097484.1 | - | 2 | 196 | |||
AK000536.1 | - | 2 | 262 | |||
AK074619.1 | - | 2 | 207 | |||
AK130791.1 | - | 2 | 204 | |||
AK302265.1 | - | 2 | 94 | |||
AK302277.1 | - | 2 | 227 | |||
AK314499.1 | - | 2 | 204 | |||
AL080162.1 | - | 2 | 271 | |||
AY358518.1 | - | 2 | 223 | |||
BC012938.1 | - | 2 | 188 | 4453977 | ||
BC047354.1 | - | 2 | 249 | |||
BX420755.2 | - | 2 | 312 | |||
KU178597.1 | - | 2 | 177 | |||
KU178598.1 | - | 2 | 177 |