Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR7C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198944.1 | NP_945182.1 | 1 | 790 | ||
XM_005259853.3 | XP_005259910.1 | 3 | 1294 | |||
XM_011527915.2 | XP_011526217.1 | 5 | 1715 | |||
GenBank mRNA | BC117212.1 | - | 1 | 865 | 40125763 | |
X89676.1 | - | 1 | 416 |