Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170753.3 | NP_736609.2 | 2 | 917 | ||
GenBank mRNA | AK074682.1 | - | 2 | 863 | ||
BC028954.1 | - | 2 | 733 | 4479639 | ||
BC063690.1 | - | 2 | 917 | 4450811 |
Gene Symbol: ERCC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277058.1 | NP_001263987.1 | 6 | 2241 | ||
NM_001277059.1 | NP_001263988.1 | 6 | 2186 | |||
GenBank mRNA | AK291018.1 | - | 6 | 2158 | ||
BC034479.1 | - | 6 | 2191 | 4822982 |