Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 7 | 722 | ||
NM_001282682.1 | NP_001269611.1 | 9 | 827 | |||
NM_001282687.1 | NP_001269616.1 | 9 | 876 | |||
NM_015677.3 | NP_056492.2 | 7 | 722 | |||
NR_104223.1 | 9 | 2281 | ||||
NR_104225.1 | 7 | 917 | ||||
NR_104226.1 | 9 | 1442 | ||||
NR_104227.1 | 9 | 1788 | ||||
GenBank mRNA | AK026507.1 | - | 7 | 657 | ||
AK026586.1 | - | 7 | 917 | |||
AK055801.1 | - | 7 | 1381 | |||
AK096440.1 | - | 9 | 1366 | |||
AK123829.1 | - | 10 | 2520 | |||
AK292312.1 | - | 9 | 771 | |||
AL050373.1 | - | 9 | 889 | |||
BC008374.1 | - | 7 | 631 | 3996066 | ||
BC008375.1 | - | 7 | 649 | 3996120 | ||
BC043234.1 | - | 9 | 2286 | |||
CR457361.1 | - | 6 | 609 |