Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015704.2 | NP_056519.1 | 5 | 541 | ||
XM_005261571.4 | XP_005261628.1 | 5 | 561 | |||
GenBank mRNA | AF038183.1 | - | 5 | 499 | ||
BC093956.1 | - | 5 | 398 | 7939801 | ||
BC112179.1 | - | 5 | 398 | 8327647 | ||
BC171724.1 | - | 3 | 306 | |||
BX403547.2 | - | 5 | 580 | |||
CR456458.1 | - | 5 | 396 |