Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPY19L2 SIRNA Assays ›
Gene Symbol:
DPY19L2
Gene Name:
dpy-19 like 2
Gene Aliases:
SPATA34, SPGF9
Chromosome Location:
Chr.12: 63558913 - 63669201 on Build GRCh38
UniGene ID:
Hs.533644
Species:
Human
siRNA ID
HSS178554
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPY19L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_173812.4
NP_776173.3
4
719
XM_006719352.2
XP_006719415.1
4
645
XM_011538215.2
XP_011536517.1
2
185
XM_017019188.1
XP_016874677.1
4
766
XM_017019189.1
XP_016874678.1
4
766
XM_017019190.1
XP_016874679.1
4
766
XM_017019191.1
XP_016874680.1
4
766
XM_017019192.1
XP_016874681.1
4
754
XM_017019193.1
XP_016874682.1
3
301
XM_017019194.1
XP_016874683.1
3
328
XM_017019195.1
XP_016874684.1
4
376
XM_017019196.1
XP_016874685.1
4
766
XM_017019197.1
XP_016874686.1
3
599
XM_017019198.1
XP_016874687.1
4
766
XM_017019199.1
XP_016874688.1
3
599
XM_017019200.1
XP_016874689.1
4
766
XM_017019201.1
XP_016874690.1
4
654
XM_017019202.1
XP_016874691.1
3
267
XM_017019203.1
XP_016874692.1
4
549
XM_017019204.1
XP_016874693.1
5
787
XM_017019205.1
XP_016874694.1
2
641
GenBank mRNA
AK057511.1
-
4
690
AK310102.1
-
3
601
AL833344.1
-
4
646
AY358792.1
-
4
640
BC068442.1
-
4
722
5297524
BC125215.1
-
4
609
BC125216.1
-
4
609
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
mannosyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0