Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303130.1 | NP_001290059.1 | 4 | 398 | ||
NM_001303131.1 | NP_001290060.1 | 4 | 629 | |||
NM_174942.2 | NP_777602.1 | 5 | 519 | |||
XM_005268804.3 | XP_005268861.1 | 6 | 588 | |||
XM_011538219.2 | XP_011536521.1 | 5 | 444 | |||
XM_011538220.2 | XP_011536522.1 | 4 | 1779 | |||
XM_011538221.2 | XP_011536523.1 | 4 | 629 | |||
XM_017019206.1 | XP_016874695.1 | 6 | 662 | |||
GenBank mRNA | AK095594.1 | - | 4 | 629 | ||
AK315189.1 | - | 4 | 376 | |||
BC043366.1 | - | 5 | 469 | 5104644 | ||
BP872524.1 | - | 4 | 398 |