Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011512262.2 | XP_011510564.1 | 10 | 1065 | ||
XM_011512265.2 | XP_011510567.1 | 8 | 544 | |||
XM_017005474.1 | XP_016860963.1 | 10 | 1065 | |||
XM_017005475.1 | XP_016860964.1 | 10 | 1065 | |||
XM_017005476.1 | XP_016860965.1 | 11 | 1095 | |||
XM_017005477.1 | XP_016860966.1 | 11 | 2690 | |||
XM_017005478.1 | XP_016860967.1 | 11 | 821 | |||
GenBank mRNA | AK092780.1 | - | 4 | 1068 | ||
BC041437.1 | - | 11 | 798 |