Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318871.1 | NP_001305800.1 | 2 | 1147 | ||
NM_207351.4 | NP_997234.3 | 2 | 1147 | |||
NR_134900.1 | 2 | 1147 | ||||
XM_011533628.2 | XP_011531930.1 | 2 | 1225 | |||
XM_011533629.2 | XP_011531931.1 | 2 | 1205 | |||
XM_017006256.1 | XP_016861745.1 | 1 | 3012 | |||
GenBank mRNA | AY358158.1 | - | 2 | 1134 | ||
BC040508.1 | - | 2 | 1126 | 5300209 | ||
BC089447.1 | - | 2 | 1106 | 30563721 | ||
BC111555.1 | - | 1 | 1044 |