Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000826.3 | NP_000817.2 | 8 | 1539 | ||
NM_001083619.1 | NP_001077088.1 | 8 | 1539 | |||
NM_001083620.1 | NP_001077089.1 | 8 | 1050 | |||
XM_017008113.1 | XP_016863602.1 | 8 | 1290 | |||
XM_017008114.1 | XP_016863603.1 | 8 | 1448 | |||
XM_017008115.1 | XP_016863604.1 | 8 | 1011 | |||
XM_017008116.1 | XP_016863605.1 | 8 | 1235 | |||
XM_017008117.1 | XP_016863606.1 | 8 | 1293 | |||
GenBank mRNA | AB209567.1 | - | 8 | 1368 | ||
AK294458.1 | - | 8 | 1216 | |||
AK308775.1 | - | 8 | 1293 | |||
AK309067.1 | - | 8 | 1063 | |||
AL832605.3 | - | 8 | 1399 | |||
BC010574.1 | - | 8 | 1359 | 4215347 | ||
BC028736.1 | - | 8 | 1028 | |||
L20814.1 | - | 8 | 1240 |