Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146156.1 | NP_001139628.1 | 9, 10 | 2079 | ||
NM_002093.3 | NP_002084.2 | 10, 11 | 2118 | |||
GenBank mRNA | AB451356.1 | - | 9, 10 | 1096 | ||
AK290897.1 | - | 9, 10 | 1452 | |||
BC000251.1 | - | 10, 11 | 1367 | 3357620 | ||
BC012760.2 | - | 9, 10 | 2078 | 3637163 | ||
CR536510.1 | - | 9, 10 | 1096 | |||
EU302497.1 | - | 9, 10 | 1096 | |||
L33801.1 | - | 9, 10 | 1135 |