Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013353.2 | NP_037485.2 | 9 | 1112 | ||
XM_011509449.1 | XP_011507751.1 | 9 | 994 | |||
XM_017001089.1 | XP_016856578.1 | 8 | 1729 | |||
XM_017001090.1 | XP_016856579.1 | 7 | 789 | |||
GenBank mRNA | AF165217.1 | - | 9 | 989 | ||
AF177173.2 | - | 9 | 1057 | |||
AK092700.1 | - | 9 | 974 | |||
AK300675.1 | - | 7 | 815 | |||
AK313280.1 | - | 9 | 1021 | |||
BX647346.1 | - | 7 | 736 |