Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198182.2 | NP_937825.2 | 9 | 1350 | ||
XM_005246159.3 | XP_005246216.1 | 8 | 1236 | |||
XM_006711882.2 | XP_006711945.1 | 9 | 1350 | |||
XM_006711884.3 | XP_006711947.1 | 8 | 1229 | |||
XM_011510343.2 | XP_011508645.1 | 9 | 1321 | |||
XM_017003900.1 | XP_016859389.1 | 9 | 1423 | |||
XM_017003901.1 | XP_016859390.1 | 9 | 1423 | |||
GenBank mRNA | AF198489.1 | - | 9 | 1288 | ||
AF411210.1 | - | 5 | 705 | |||
AK293855.1 | - | 9 | 1397 | |||
AK312950.1 | - | 9 | 1320 | |||
BC067520.1 | - | 9 | 1231 | 6971724 |