Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324458.1 | NP_001311387.1 | 7 | 533 | ||
NM_001324459.1 | NP_001311388.1 | 7 | 561 | |||
NM_013292.4 | NP_037424.2 | 5 | 396 | |||
GenBank mRNA | AF363061.1 | - | 5 | 369 | ||
AK314082.1 | - | 5 | 396 | |||
BC012571.1 | - | 5 | 374 | 4245958 | ||
BI828249.1 | - | 7 | 561 | |||
BI837593.1 | - | 7 | 538 | |||
BX649054.1 | - | 5 | 364 | |||
CR456963.1 | - | 4 | 315 | |||
M21812.1 | - | 5 | 376 | |||
X57543.1 | - | 5 | 376 |