Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013335.3 | NP_037467.2 | 8, 9 | 839 | ||
NM_205847.2 | NP_995319.1 | 8, 9 | 1120 | |||
XM_005246483.3 | XP_005246540.1 | 9, 10 | 854 | |||
XM_005246486.3 | XP_005246543.1 | 8, 9 | 808 | |||
XM_011511032.2 | XP_011509334.1 | 9, 10 | 823 | |||
XM_017003906.1 | XP_016859395.1 | 9, 10 | 854 | |||
XM_017003907.1 | XP_016859396.1 | 9, 10 | 823 | |||
XM_017003908.1 | XP_016859397.1 | 9, 10 | 1129 | |||
XM_017003909.1 | XP_016859398.1 | 8, 9 | 839 | |||
XM_017003910.1 | XP_016859399.1 | 8, 9 | 808 | |||
XM_017003911.1 | XP_016859400.1 | 8, 9 | 1114 | |||
XM_017003914.1 | XP_016859403.1 | 9, 10 | 783 | |||
XM_017003915.1 | XP_016859404.1 | 9, 10 | 1013 | |||
XM_017003916.1 | XP_016859405.1 | 8, 9 | 768 | |||
XM_017003917.1 | XP_016859406.1 | 8, 9 | 768 | |||
GenBank mRNA | AF135422.2 | - | 7, 8 | 818 | ||
AK000999.1 | - | 8, 9 | 813 | |||
AK022578.1 | - | 8, 9 | 1114 | |||
AK222951.1 | - | 8, 9 | 803 | |||
AK290671.1 | - | 8, 9 | 1116 | |||
BC007456.2 | - | 8, 9 | 1108 | 2989461 |