Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308187.1 | NP_001295116.1 | 3 | 332 | ||
NM_001308190.1 | NP_001295119.1 | 2 | 233 | |||
NM_002099.7 | NP_002090.4 | 3 | 332 | |||
XM_017008134.1 | XP_016863623.1 | 3 | 336 | |||
XM_017008135.1 | XP_016863624.1 | 4 | 399 | |||
XM_017008136.1 | XP_016863625.1 | 2 | 301 | |||
GenBank mRNA | AK290561.1 | - | 3 | 297 | ||
AY950614.1 | - | 3 | 181 | |||
BC005319.1 | - | 3 | 238 | 3930700 | ||
BC013328.1 | - | 3 | 219 | 4134368 | ||
DA537533.1 | - | 3 | 297 | |||
DA539682.1 | - | 3 | 333 | |||
EU338231.1 | - | 4 | 301 | |||
EU338232.1 | - | 3 | 218 | |||
EU338233.1 | - | 4 | 301 | |||
EU338234.1 | - | 2 | 125 | |||
GU347002.1 | - | 4 | 325 | |||
GU347003.1 | - | 4 | 404 | |||
GU347004.1 | - | 4 | 382 | |||
GU347006.1 | - | 3 | 224 | |||
GU347008.1 | - | 2 | 119 | |||
L31860.1 | - | 3 | 236 | |||
M12857.1 | - | 3 | 191 | |||
M36281.1 | - | 3 | 191 | |||
U00177.1 | - | 3 | 181 | |||
X08054.1 | - | 3 | 237 | |||
X51798.1 | - | 3 | 205 | |||
X57023.1 | - | 3 | 181 |