Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022075.4 | NP_071358.1 | 10 | 1342 | ||
NM_181746.3 | NP_859530.1 | 10 | 1382 | |||
XM_011509451.2 | XP_011507753.1 | 10 | 1045 | |||
XM_011509452.2 | XP_011507754.1 | 10 | 1093 | |||
GenBank mRNA | AF177338.2 | - | 10 | 1071 | ||
AF189062.4 | - | 5 | 555 | |||
AK001105.1 | - | 9 | 1011 | |||
AK290989.1 | - | 10 | 1018 | |||
AY091458.1 | - | 10 | 1164 | |||
BC001357.2 | - | 10 | 1037 | 3051086 | ||
BC010032.2 | - | 10 | 1001 | 3506408 |