Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256588.1 | NP_001243517.1 | 1 | 235 | ||
NM_014579.3 | NP_055394.2 | 1 | 235 | |||
GenBank mRNA | AF186081.1 | - | 1 | 179 | ||
AK314974.1 | - | 1 | 199 | |||
BC096723.1 | - | 1 | 66 | 40004409 | ||
BC110057.1 | - | 1 | 66 | 40004405 | ||
BP359281.1 | - | 1 | 237 | |||
CN306082.1 | - | 1 | 161 | |||
D83039.1 | - | 1 | 173 | |||
DA871745.1 | - | 1 | 199 |