Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005317.3 | NP_005308.1 | 1, 2 | 91 | ||
GenBank mRNA | BC025701.1 | - | 1, 2 | 63 | 5222281 | |
L23134.1 | - | 1, 2 | 53 |