Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037811.2 | NP_001032900.1 | 2 | 195 | ||
NM_004493.2 | NP_004484.1 | 2 | 195 | |||
GenBank mRNA | AF035555.2 | - | 2 | 200 | ||
AF069134.1 | - | 2 | 182 | |||
AU132847.1 | - | 2 | 195 | |||
AY092415.1 | - | 1 | 14 | |||
BC000372.2 | - | 2 | 179 | 2819721 | ||
BC000829.1 | - | 2 | 179 | |||
BC008708.1 | - | 2 | 175 | 3160844 | ||
CR456723.1 | - | 2 | 164 | |||
KU178107.1 | - | 2 | 164 | |||
KU178108.1 | - | 2 | 164 | |||
U73514.1 | - | 2 | 179 | |||
U96132.1 | - | 2 | 191 |