Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040427.1 | NP_001035517.1 | 7 | 1083 | ||
NM_001286249.1 | NP_001273178.1 | 5 | 878 | |||
NM_005326.4 | NP_005317.2 | 6 | 987 | |||
XM_011522469.2 | XP_011520771.1 | 6 | 627 | |||
XM_011522470.2 | XP_011520772.1 | 5 | 516 | |||
XM_017023195.1 | XP_016878684.1 | 6 | 552 | |||
GenBank mRNA | AK290155.1 | - | 6 | 552 | ||
AK298174.1 | - | 5 | 518 | |||
AK299173.1 | - | 6 | 618 | |||
AK299993.1 | - | 6 | 618 | |||
BC000840.1 | - | 6 | 607 | 3457214 | ||
BC002627.2 | - | 6 | 936 | 3608098 | ||
BU176541.1 | - | 7 | 689 | |||
X90999.1 | - | 5 | 472 |