Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312672.1 | NP_001299601.1 | 2 | 306 | ||
NM_005666.3 | NP_005657.1 | 4 | 678 | |||
XM_005245113.3 | XP_005245170.1 | 3 | 497 | |||
XM_011509458.2 | XP_011507760.1 | 4 | 650 | |||
XM_011509459.2 | XP_011507761.1 | 4 | 712 | |||
XM_011509460.2 | XP_011507762.1 | 3 | 487 | |||
XM_017001109.1 | XP_016856598.1 | 3 | 485 | |||
GenBank mRNA | AX772812.1 | - | 2 | 216 | ||
BC022283.1 | - | 4 | 667 | 4734331 | ||
BG563114.1 | - | 4 | 676 | |||
BG566607.1 | - | 2 | 294 | |||
X64877.1 | - | 4 | 642 |