Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNIP2 SIRNA Assays ›
Gene Symbol:
KCNIP2
Gene Name:
potassium voltage-gated channel interacting protein 2
Gene Aliases:
KCHIP2
Chromosome Location:
Chr.10: 101825971 - 101843920 on Build GRCh38
UniGene ID:
Hs.97044
Species:
Human
siRNA ID
HSS179211
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNIP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014591.4
NP_055406.2
4
715
NM_173191.2
NP_775283.1
4
670
NM_173192.2
NP_775284.1
3
616
NM_173193.2
NP_775285.1
2
520
NM_173194.2
NP_775286.1
2
197
NM_173195.2
NP_775287.1
2
520
NM_173197.2
NP_775289.1
2
520
XM_005269729.2
XP_005269786.1
3
518
XM_005269730.2
XP_005269787.1
2
475
XM_006717812.2
XP_006717875.1
4
572
XM_011539731.2
XP_011538033.1
2
212
XM_017016161.1
XP_016871650.1
2
267
GenBank mRNA
AF199598.1
-
3
264
AF295076.1
-
2
168
AF295530.1
-
2
168
AF347114.1
-
2
168
AF367018.1
-
4
363
AF367019.1
-
2
168
AF367020.1
-
1
110
AF367021.1
-
4
318
AK027347.1
-
2
188
AK055089.1
-
4
594
AK094668.1
-
5
694
AK125382.1
-
2
1227
AK301433.1
-
2
189
AK309368.1
-
2
396
AK314536.1
-
2
401
AK315042.1
-
3
496
AL136722.1
-
2
215
AY026328.1
-
2
168
AY302141.1
-
2
168
BC034685.1
-
3
479
4155871
DQ148480.1
-
4
318
DQ148481.1
-
3
264
DQ148482.1
-
2
168
DQ148483.1
-
2
183
DQ148484.1
-
1
110
DQ862463.1
-
2
168
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
detection of calcium ion
potassium ion transport
muscle contraction
signal transduction
chemical synaptic transmission
regulation of heart contraction
clustering of voltage-gated potassium channels
cardiac conduction
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
regulation of cation channel activity
A-type (transient outward) potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
ion channel binding
ER retention sequence binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0