Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014601.3 | NP_055416.2 | 2 | 538 | ||
GenBank mRNA | AF454952.1 | - | 1 | 343 | ||
AK074701.1 | - | 2 | 460 | |||
AK315548.1 | - | 2 | 462 | |||
BC014445.1 | - | 2 | 449 | 4908085 | ||
DA094880.1 | - | 2 | 540 |