Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243961.1 | NP_001230890.1 | 2, 3 | 451 | ||
NM_002123.4 | NP_002114.3 | 2, 3 | 451 | |||
GenBank mRNA | AF000447.1 | - | 2, 3 | 362 | ||
AK295695.1 | - | 2, 3 | 419 | |||
AK296613.1 | - | 2, 3 | 430 | |||
AK316333.1 | - | 2, 3 | 443 | |||
BC128595.1 | - | 1, 2 | 265 | |||
DC409119.1 | - | 2, 3 | 451 | |||
L34104.1 | - | 1, 2 | 273 | |||
L34105.1 | - | 1, 2 | 273 | |||
L34106.1 | - | 1, 2 | 273 | |||
L40181.1 | - | 1, 2 | 273 | |||
M16276.1 | - | 2, 3 | 579 | |||
M20432.1 | - | 2, 3 | 428 | |||
M57649.1 | - | 2, 3 | 355 | |||
M57650.1 | - | 1, 2 | 115 | |||
M59801.1 | - | 1, 2 | 273 |