Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175066.3 | NP_778236.2 | 9 | 1327 | ||
XM_011538256.1 | XP_011536558.1 | 9 | 1327 | |||
GenBank mRNA | AB593157.1 | - | 9 | 1347 | ||
AK097078.1 | - | 9 | 1327 | |||
BC040185.2 | - | 9 | 1300 | 4801253 | ||
CR936870.1 | - | 1 | 346 |