Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300868.1 | NP_001287797.1 | 9 | 1094 | ||
NM_001300869.1 | NP_001287798.1 | 9 | 1059 | |||
NM_001532.2 | NP_001523.2 | 7 | 893 | |||
NR_125343.1 | 7 | 893 | ||||
XM_011544974.1 | XP_011543276.1 | 9 | 1044 | |||
XM_017017630.1 | XP_016873119.1 | 7 | 1247 | |||
XM_017017631.1 | XP_016873120.1 | 7 | 1247 | |||
XM_017017632.1 | XP_016873121.1 | 9 | 1044 | |||
XM_017017633.1 | XP_016873122.1 | 7 | 1247 | |||
XM_017017634.1 | XP_016873123.1 | 7 | 1247 | |||
XM_017017635.1 | XP_016873124.1 | 7 | 1247 | |||
XM_017017636.1 | XP_016873125.1 | 7 | 1247 | |||
XM_017017637.1 | XP_016873126.1 | 7 | 1247 | |||
XM_017017638.1 | XP_016873127.1 | 5 | 417 | |||
GenBank mRNA | AF029358.1 | - | 7 | 667 | ||
AF034102.1 | - | 7 | 901 | |||
AF401235.1 | - | 7 | 901 | |||
AK057041.1 | - | 8 | 884 | |||
BC011387.1 | - | 8 | 774 | 3872824 | ||
BC093634.1 | - | 7 | 713 | 7939479 | ||
BC143335.1 | - | 7 | 713 | |||
X86681.1 | - | 7 | 659 |