Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000383.3 | NP_000374.1 | 10 | 1336 | ||
GenBank mRNA | AB006682.1 | - | 10 | 1294 | ||
AB006683.1 | - | 4 | 812 | |||
AK309572.1 | - | 10 | 1898 | |||
BC137268.1 | - | 3 | 993 | 9021265 | ||
BC137270.1 | - | 3 | 992 | 9021267 | ||
Z97990.1 | - | 10 | 1287 |