Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002925.1 | NP_001002925.1 | 1 | 102 | ||
GenBank mRNA | BC136989.1 | - | 1 | 134 | 9020981 | |
BC136990.1 | - | 1 | 134 | 9020982 |