Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198317.2 | NP_938073.1 | 12 | 1927 | ||
XM_006710600.3 | XP_006710663.1 | 12 | 2480 | |||
GenBank mRNA | AK127585.1 | - | 11 | 1574 | ||
AY423763.1 | - | 12 | 1927 | |||
BC033192.1 | - | 5 | 616 | |||
BC105742.1 | - | 9 | 1430 | |||
CA432461.1 | - | 1 | 647 |