Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZBTB8OS SIRNA Assays ›
Gene Symbol:
ZBTB8OS
Gene Name:
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases:
ARCH, ARCH2
Chromosome Location:
Chr.1: 32620820 - 32651013 on Build GRCh38
UniGene ID:
Hs.655921
Species:
Human
siRNA ID
HSS179516
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZBTB8OS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308135.1
NP_001295064.1
6
506
NM_001308136.1
NP_001295065.1
6
474
NM_001308137.1
NP_001295066.1
5
453
NM_001308138.1
NP_001295067.1
6
571
NM_001308139.1
NP_001295068.1
7
565
NM_001308140.1
NP_001295069.1
7
558
NM_001308141.1
NP_001295070.1
7
531
NM_178547.3
NP_848642.1
7
527
XM_011541328.1
XP_011539630.1
7
552
XM_011541329.2
XP_011539631.1
7
452
XM_011541330.2
XP_011539632.1
6
938
XM_011541334.2
XP_011539636.1
6
897
XM_011541335.2
XP_011539637.1
7
504
XM_011541337.2
XP_011539639.1
7
539
XM_011541338.2
XP_011539640.1
6
898
XM_017001134.1
XP_016856623.1
7
538
XM_017001136.1
XP_016856625.1
5
749
XM_017001137.1
XP_016856626.1
7
386
XM_017001138.1
XP_016856627.1
6
424
GenBank mRNA
AK289480.1
-
7
869
AY099494.1
-
7
427
AY151084.1
-
7
521
BC058843.1
-
6
444
6745279
BE780045.1
-
6
467
BF216169.1
-
7
505
BM465977.1
-
2
159
CK819207.1
-
1
81
CN344731.1
-
6
414
DT220608.1
-
6
531
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
tRNA splicing, via endonucleolytic cleavage and ligation
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0