Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005473.2 | NP_001005473.1 | 2 | 777 | ||
XM_017009436.1 | XP_016864925.1 | 2 | 699 | |||
XM_017009437.1 | XP_016864926.1 | 2 | 1497 | |||
XM_017009438.1 | XP_016864927.1 | 2 | 805 | |||
GenBank mRNA | AK127142.1 | - | 2 | 750 | ||
BX648329.1 | - | 2 | 778 |