Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 5 | 768 | ||
NM_001197079.1 | NP_001184008.1 | 4 | 658 | |||
NM_001197080.1 | NP_001184009.1 | 4 | 493 | |||
NM_001550.3 | NP_001541.2 | 4 | 559 | |||
NR_120333.1 | 4 | 479 | ||||
XM_011516142.2 | XP_011514444.1 | 4 | 554 | |||
XM_011516143.2 | XP_011514445.1 | 4 | 739 | |||
XM_017012151.1 | XP_016867640.1 | 4 | 806 | |||
GenBank mRNA | AK225222.1 | - | 4 | 532 | ||
AK297509.1 | - | 4 | 304 | |||
AK298894.1 | - | 4 | 299 | |||
AK308066.1 | - | 5 | 743 | |||
AK313211.1 | - | 5 | 768 | |||
AK315967.1 | - | 4 | 491 | |||
AU280361.1 | - | 4 | 556 | |||
BC001272.1 | - | 4 | 512 | 3458309 | ||
BT019889.1 | - | 4 | 298 | |||
BT019890.1 | - | 4 | 298 |