Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286509.1 | NP_001273438.1 | 4 | 1163 | ||
NM_001286510.1 | NP_001273439.1 | 4 | 1162 | |||
NM_001286511.1 | NP_001273440.1 | 4 | 1117 | |||
NM_001286512.1 | NP_001273441.1 | 4 | 1166 | |||
NM_001286513.1 | NP_001273442.1 | 3 | 1012 | |||
NM_001286517.1 | NP_001273446.1 | 3 | 973 | |||
NM_001286519.1 | NP_001273448.1 | 2 | 818 | |||
NM_001286520.1 | NP_001273449.1 | 4 | 1171 | |||
NM_178148.3 | NP_835361.1 | 4 | 1167 | |||
GenBank mRNA | AB097021.1 | - | 5 | 1004 | ||
AB097039.1 | - | 5 | 1004 | |||
AB106538.1 | - | 4 | 1015 | |||
AK123749.1 | - | 4 | 1064 | |||
AK223028.1 | - | 3 | 817 | |||
AK293251.1 | - | 2 | 785 | |||
AK293344.1 | - | 3 | 969 | |||
AK307572.1 | - | 4 | 1102 | |||
AY491520.1 | - | 4 | 1133 | |||
BC016839.1 | - | 1 | 190 | |||
BC024288.1 | - | 4 | 1131 | 4890949 |