Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12RB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258214.1 | NP_001245143.1 | 7 | 1418 | ||
NM_001258215.1 | NP_001245144.1 | 6 | 1330 | |||
NM_001258216.1 | NP_001245145.1 | 6 | 1330 | |||
NM_001319233.1 | NP_001306162.1 | 7 | 1414 | |||
NM_001559.2 | NP_001550.1 | 6 | 1330 | |||
NR_047583.1 | 6 | 1330 | ||||
NR_047584.1 | 6 | 1330 | ||||
XM_005270825.2 | XP_005270882.1 | 7 | 1431 | |||
XM_005270827.2 | XP_005270884.1 | 7 | 1427 | |||
XM_005270828.3 | XP_005270885.1 | 6 | 1339 | |||
XM_006710617.2 | XP_006710680.1 | 7 | 1431 | |||
XM_011541383.2 | XP_011539685.1 | 7 | 883 | |||
XM_011541384.2 | XP_011539686.1 | 7 | 1431 | |||
XM_017001202.1 | XP_016856691.1 | 7 | 1431 | |||
XM_017001203.1 | XP_016856692.1 | 6 | 1343 | |||
XM_017001204.1 | XP_016856693.1 | 7 | 1431 | |||
GenBank mRNA | AK294494.1 | - | 7 | 909 | ||
AK316525.1 | - | 7 | 889 | |||
BC104772.1 | - | 5 | 740 | 8143775 | ||
BC104774.1 | - | 5 | 740 | 8143777 | ||
BC143248.1 | - | 5 | 740 | |||
BC143249.1 | - | 5 | 740 | 9051751 | ||
BC143250.1 | - | 5 | 740 | |||
U64198.1 | - | 6 | 1330 |