Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267728.1 | NP_001254657.1 | 2 | 421 | ||
NM_005537.5 | NP_005528.4 | 2 | 1326 | |||
NM_198217.2 | NP_937860.1 | 2 | 684 | |||
NM_198218.2 | NP_937861.1 | 2 | 653 | |||
NM_198219.2 | NP_937862.1 | 2 | 1352 | |||
GenBank mRNA | AB024401.1 | - | 2 | 915 | ||
AB024402.1 | - | 2 | 1355 | |||
AB031269.1 | - | 2 | 770 | |||
AB037594.2 | - | 2 | 421 | |||
AF001954.1 | - | 2 | 525 | |||
AF044076.1 | - | 2 | 509 | |||
AF078835.1 | - | 2 | 465 | |||
AF149721.1 | - | 2 | 915 | |||
AF149722.1 | - | 2 | 456 | |||
AF149723.1 | - | 2 | 571 | |||
AF181849.1 | - | 2 | 1326 | |||
AF181850.1 | - | 2 | 1337 | |||
AJ310392.1 | - | 2 | 470 | |||
AK302353.1 | - | 2 | 684 | |||
BC018348.1 | - | 2 | 761 | |||
BC093942.1 | - | 2 | 517 | 7939787 | ||
BC093944.1 | - | 2 | 517 | 7939789 | ||
BG703874.1 | - | 2 | 658 | |||
BQ430264.1 | - | 2 | 361 |