Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321042.1 | NP_001307971.1 | 11 | 1476 | ||
NM_005539.4 | NP_005530.3 | 14 | 1423 | |||
XM_017016204.1 | XP_016871693.1 | 13 | 1077 | |||
GenBank mRNA | BC052265.1 | - | 11 | 850 | ||
BC062300.1 | - | 11 | 850 | |||
BC096280.2 | - | 15 | 1167 | 40006844 | ||
BC096281.2 | - | 14 | 1086 | 40006846 | ||
BC117515.1 | - | 1, 2 | 102 | |||
X77567.1 | - | 14 | 1204 | |||
Z31695.1 | - | 14 | 1059 |