Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324237.1 | NP_001311166.1 | 2 | 179 | ||
NM_001324240.1 | NP_001311169.1 | 2 | 271 | |||
NM_033453.3 | NP_258412.1 | 2 | 271 | |||
NM_181493.3 | NP_852470.1 | 2 | 220 | |||
NR_052000.1 | 2 | 179 | ||||
NR_052002.1 | 2 | 271 | ||||
XM_006723564.3 | XP_006723627.1 | 2 | 554 | |||
XM_011529234.2 | XP_011527536.1 | 2 | 554 | |||
GenBank mRNA | AB062127.1 | - | 2 | 140 | ||
AF026816.2 | - | 2 | 130 | |||
AF063607.1 | - | 2 | 155 | |||
AF219116.1 | - | 2 | 183 | |||
BC010138.2 | - | 2 | 115 | 4124624 | ||
BF969192.1 | - | 2 | 179 | |||
BM560812.1 | - | 271 | ||||
BQ049090.1 | - | 2 | 118 | |||
EF199841.1 | - | 2 | 118 | |||
EF213026.1 | - | 2 | 67 | |||
KU178148.1 | - | 2 | 79 |