Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002226.4 | NP_002217.3 | 25 | 3625 | ||
NM_145159.2 | NP_660142.1 | 24 | 3511 | |||
GenBank mRNA | AB209370.1 | - | 1 | 3643 | ||
AF003521.1 | - | 25 | 3253 | |||
AF020201.1 | - | 25 | 3272 | |||
AF029778.1 | - | 25 | 3625 | |||
AF029779.1 | - | 24 | 3511 | |||
Y14330.1 | - | 25 | 3176 |