Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175078.2 | NP_778253.2 | 1 | 202 | ||
XM_011538289.2 | XP_011536591.1 | 1 | 263 | |||
GenBank mRNA | AJ564104.1 | - | 1 | 202 | ||
BC118598.1 | - | 1 | 202 | 40108170 | ||
BC122558.1 | - | 1 | 202 | 40108176 |