Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164664.1 | NP_001158136.1 | 26 | 3941 | ||
NM_001290226.1 | NP_001277155.1 | 24 | 3393 | |||
NM_001290227.1 | NP_001277156.1 | 23 | 2964 | |||
NM_001297651.1 | NP_001284580.1 | 23 | 3425 | |||
NM_015183.2 | NP_055998.1 | 25 | 3134 | |||
XM_006714606.3 | XP_006714669.1 | 24 | 3387 | |||
XM_006714610.2 | XP_006714673.1 | 22 | 3173 | |||
XM_011543382.2 | XP_011541684.1 | 28 | 4033 | |||
XM_011543384.2 | XP_011541686.1 | 23 | 5092 | |||
XM_011543385.2 | XP_011541687.1 | 23 | 6212 | |||
XM_011543386.2 | XP_011541688.1 | 23 | 3043 | |||
XM_017009447.1 | XP_016864936.1 | 29 | 4114 | |||
XM_017009448.1 | XP_016864937.1 | 27 | 3997 | |||
XM_017009449.1 | XP_016864938.1 | 24 | 3432 | |||
XM_017009450.1 | XP_016864939.1 | 24 | 3126 | |||
XM_017009451.1 | XP_016864940.1 | 23 | 2925 | |||
XM_017009452.1 | XP_016864941.1 | 24 | 6441 | |||
XM_017009453.1 | XP_016864942.1 | 23 | 13256 | |||
GenBank mRNA | AB002301.1 | - | 15 | 2177 | ||
AK296707.1 | - | 23 | 2929 | |||
AY830839.1 | - | 25 | 3135 | |||
BX648980.1 | - | 5 | 743 |